Learn More
Hoefer™ Divider Plates for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units
SDP
Catalog No. 03500205
Click to view available options
For Use With (Equipment):
SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit
Catalog No. For Use With (Equipment)
03-500-205 SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit
1 options
Catalog No. 03-500-205 Supplier Hoefer™ Supplier No. SE6102D
Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units
  • Notched
  • 18 x 16 cm
  • Glass
Specifications
For Use With (Equipment) SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit
Type Notched Glass Plate Divider
Material Glass