Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ GD 2000 Vacuum Gel Dryer Accessories: Rubber Overlay Sheet
SDP

Catalog No. 03500453
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
GD 2000 Vacuum Gel Dryer System, SE 1140, SE 1150, SE 1160 and SE 1175 Gel Dryers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
03-500-453 GD 2000 Vacuum Gel Dryer System, SE 1140, SE 1150, SE 1160 and SE 1175 Gel Dryers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-500-453 Supplier Hoefer™ Supplier No. SE1143
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dries acrylamide and agarose gels rapidly, evenly and safely.

  • Clear Silicone Rubber Overlay Sheet

Specifications

Color Clear
For Use With (Equipment) GD 2000 Vacuum Gel Dryer System, SE 1140, SE 1150, SE 1160 and SE 1175 Gel Dryers
Type Rubber Overlay Sheet
Material Silicone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.