Learn More
Hoefer™ Gel Casting Tray for SUB25 Maxi-Plus
SDP
Catalog No. 03500800
Click to view available options
For Use With (Equipment):
SUB25 Maxi-Plus unit
Catalog No. For Use With (Equipment)
03-500-800 SUB25 Maxi-Plus unit
1 options
Catalog No. 03-500-800 Supplier Hoefer™ Supplier No. SUB25UT
For use with SUB25 and SUB25C Submarine Gel Electrophoresis Units
  • 25 x 30 cm
  • Casting Gates
Specifications
Description Gel Casting Tray
For Use With (Equipment) SUB25 Maxi-Plus unit
Dimensions (L x W) 25 x 30 cm