Learn More
Hoefer™ Gel Plate Rack
SDP
Catalog No. SE914
Click to view available options
For Use With (Equipment):
SE 900 Large format Vertical Electrophoresis System
Catalog No. For Use With (Equipment)
SE914 SE 900 Large format Vertical Electrophoresis System
1 options
Catalog No. SE914 Supplier Hoefer™ Supplier No. SE914
Compatible with SE 900 Vertical Electrophoresis System
Holds up to 14 large-format gel cassettes.
Specifications
Device Holds Up to 14 Large Format Gel Cassette
For Use With (Equipment) SE 900 Large format Vertical Electrophoresis System
Type Gel Plate Rack