Learn More
Hoefer™ Gel Seal
SDP
Catalog No. 03500255
Click to view available options
Volume (English):
0.25 oz.
Catalog No. Volume (English)
03-500-255 0.25 oz.
1 options
Catalog No. 03-500-255 Supplier Hoefer™ Supplier No. SE6070
A sealant used very sparingly in vertical gel casting to reduce leaking
  • For use with E245 Dual Gel Caster
Specifications
Type Gel Seal
Volume (English) 0.25 oz.