Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ HE 33 Mini Submarine Electrophoresis Unit
SDP

Catalog No. 03500130
Encompass
Encompass_Preferred
Change view
Click to view available options
Model:
Hoefer HE 33 Complete
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
03-500-130 Hoefer HE 33 Complete
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-500-130 Supplier Hoefer™ Supplier No. HE3381.5
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Buffer chamber, lid with cables, UV-transparent gel running tray, gel casting tray, 4 foam gaskets, bubble level. Complete system (03-500-130) also includes 1.5mm thick 8-well comb and comb back.

Designed for very fast separations of DNA restriction fragments in agarose gels

  • The clear molded base acts as a heat-sink—simply fill the base with coolant and seal
  • Cooling the unit prior to a run allows higher voltages to be applied without generating band distortions or damaging the gel and unit
  • No taping required for casting
  • Electrical safety locks in lid

Specifications

Certifications/Compliance EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
Gel Size Mini
Max. Current 500 mA
Type Mini Horizontal Agarose Electrophoresis Unit
Humidity Range Up to 80%
Includes Buffer chamber assembly, safety lid with high voltage Leads, running tray, casting tray, Foam gaskets, bubble level, 1.5mm thick, 8-well comb, comb back
Model Hoefer HE 33 Complete
Sample Size Up to 32
Temperature (English) Maximum 122°F
Temperature (Metric) Maximum 50°C
Volume (English) Buffer 8.4 oz.
Volume (Metric) Buffer 250 mL
Dimensions (L x W x H) 9.4 x 5.1 x 2.8 in. (24 x 13 x 7 cm)
Length (English) 9.4 in.
Length (English) Gel 3.9 in.
Length (Metric) 24 cm
Length (Metric) Gel 10 cm
Width (English) 5.1 in.
Width (English) Gel 2.7 in.
Width (Metric) 13 cm
Width (Metric) Gel 7 cm
Height (English) 2.8 in.
Height (Metric) 7 cm
Voltage 500 V
Wattage 15 w
Amperage 500 mA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.