Learn More
Hoefer™ miniVE Electrophoresis Unit Accessories and Parts
SDP
Catalog No. p-3314751
Click to view available options
Type:
Banana Plug Kit
Clamp Screw
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
03-500-499 Banana Plug Kit
03-500-496 Clamp Screw
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-500-499 Supplier Hoefer™ Supplier No. SE314
Only null left
Add to Cart
Add to Cart
Part and accessories for miniVE Electrophoresis Unit
  • Accesories include High Voltage Leads (2 pcs), Banana Plug Kit (2 plugs, 2 nuts, 4 washers), Foam Gasket, Clamp Screws (5 pcs), Upper Buffer Chamber Assembly, Clamp Assembly, Sealing Plate Assembly, Sealing Gasket and Lower Buffer Chamber (tank)
Specifications
For Use With (Equipment) MiniVE Vertical Electrophoresis System
Type Banana Plug Kit
Includes 2 Plugs, 2 nuts and 4 washers