Learn More
Hoefer™ Plate for SE 640 Vertical Electrophoresis Unit
SDP
Catalog No. SE6402D
Click to view available options
For Use With (Equipment):
SE 640 Vertical Electrophoresis Unit
Catalog No. For Use With (Equipment)
SE6402D SE 640 Vertical Electrophoresis Unit
1 options
Catalog No. SE6402D Supplier Hoefer™ Supplier No. SE6402D
For use with SE660, SE600X Chroma, SE600, SE640, SE410 and SE400
  • Notched
  • 18 x 8 cm
  • Glass
Specifications
For Use With (Equipment) SE 640 Vertical Electrophoresis Unit
Type Notched Glass Plate Divider
Material Glass