Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ PlateMate™ Washing System Accessories: Plate Adapter
SDP

Catalog No. 03500449
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
SE 100 Platemate™ Plate Washing System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
03-500-449 SE 100 Platemate™ Plate Washing System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-500-449 Supplier Hoefer™ Supplier No. SE103
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adapter for PlateMate Washing System

For 8, 10.5, and 12cm plates.

Specifications

Device Holds 8, 10.5 and 12 cm Plates
For Use With (Equipment) SE 100 Platemate™ Plate Washing System
Type Long Plate Adapter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.