Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ SG Gradient Maker: Outlet Fitting
SDP

Catalog No. 03500275
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
SG 5, SG 15, SG 30, SG 50 or SG 100 Gradient Maker
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
03-500-275 SG 5, SG 15, SG 30, SG 50 or SG 100 Gradient Maker
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-500-275 Supplier Hoefer™ Supplier No. SG1006
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Outlet fitting for SG Gradient Makers

For use with SG Gradient Makers

Specifications

Color White
For Use With (Equipment) SG 5, SG 15, SG 30, SG 50 or SG 100 Gradient Maker
Type Luer Outlet Fitting
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.