Learn More
Hoefer™ Upper Buffer Chamber for SE 400 Vertical Electrophoresis Unit
SDP
Catalog No. 03500234
Click to view available options
For Use With (Equipment):
SE 400 Vertical Electrophoresis Unit
Catalog No. For Use With (Equipment)
03-500-234 SE 400 Vertical Electrophoresis Unit
1 options
Catalog No. 03-500-234 Supplier Hoefer™ Supplier No. SE4154
Includes
Slotted gasket
Upper Buffer Chamber for SE 400 Vertical Electrophoresis Unit
  • For use with SE 400 Vertical Electrophoresis Unit
Specifications
For Use With (Equipment) SE 400 Vertical Electrophoresis Unit
Type Upper Buffer Chamber
Includes Slotted gasket