Description
- Comfortable - High-density nitrile-blended cushion helps eliminate pressure to the lower back and legs, providing superior comfort
- Safe - Certified high-traction by the National Floor Safety Institute (NFSI); edges will not crack or curl and provide a safe transition from floor to mat
- Durable - Nitrile-blended cushion provides years of useful service life
- Cushion is molded, not glued, to the nitrile rubber surface so it will not separate
- Versatile - Welding safe, grease/oil resistant, chemical resistant, ESD rating of electrically conductive
- Available in 5/8 in. or 7/8 in. thicknesses with solid black or with OSHA approved yellow striped borders
- Can be customized with your logo, design, or message
Specifications
Specifications
| Certifications/Compliance | NFSI Certified. Made in USA |
| Dimensions (L x W) | 2 x 12 ft. |
| Thickness (English) | 5/8 in. |
| Color | Black |