Description
- Comfortable - High-density, closed-cell nitrile rubber cushion provides excellent antifatigue qualities to reduce the pain and fatigue associated with standing for long periods of time
- Outstanding anti-fatigue qualities and perform in various environments including those with oil, grease, and chemicals.
- Safe - Certified high-traction by the National Floor Safety Institute (NFSI); beveled edges provide a safe transition from floor to mat and will not crack or curl
- Durable - Nitrile-blended cushion provides years of useful life and is molded, not glued, to the nitrile rubber surface so it will not separate
- Versatile - Welding safe, grease/oil resistant, chemical resistant, ESD rating of electrically conductive
- Mats and tiles are 3/4 in. thick and available with black or OSHA-approved yellow striped border
Specifications
Specifications
| Style | Yellow/Black-striped border |
| Type | Side Tile |
| Certifications/Compliance | NFSI Certified. Made in USA |
| Dimensions (L x W) | 39.875 x 39.875 in. |
| Thickness (English) | 5/8 in. |
| Color | Black |