Learn More
M+A Matting Hog Heaven™ Prime Anti-Fatigue Mat
Catalog No. 17555101 Shop All M+A Matting Products
Change view
Click to view available options
Dimensions (L x W):
3 x 5 ft.
3 x 10 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17-555-101 3 x 5 ft.
17-555-102 3 x 10 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-555-101 Supplier M+A Matting Supplier No. 4222000035
Hog Heaven™ Anti-Fatigue Mats are designed to provide comfort and durability in a variety of harsh conditions.
  • Comfortable - High-density nitrile-blended cushion helps eliminate pressure to the lower back and legs, providing superior comfort
  • Safe - Certified high-traction by the National Floor Safety Institute (NFSI); edges will not crack or curl and provide a safe transition from floor to mat
  • Durable - Nitrile-blended cushion provides years of useful service life
  • Cushion is molded, not glued, to the nitrile rubber surface so it will not separate
  • Versatile - Welding safe, grease/oil resistant, chemical resistant, ESD rating of electrically conductive
  • Available in 5/8 in. or 7/8 in. thicknesses with solid black or with OSHA approved yellow striped borders
  • Can be customized with your logo, design, or message
Specifications
Certifications/Compliance NFSI Certified. Made in USA
Color Black
Dimensions (L x W) 3 x 5 ft.
Thickness (English) 5/8 in.