Learn More
Description
Kit Contents:
- hOGG1 (100 units)
- 25X FLARE™ Buffer 1 (40 mL)
- REC™Dilution Buffer (10 mL)
- 100X BSA Additive (100 μL)
Specifications
Specifications
| Content And Storage | Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| No. of Reactions | 100 Tests |
| Species | Human |
| Target | OGG1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
