Learn More
JDM SCIENTIFIC LLC Holder for chromatography column
Supplier:  JDM SCIENTIFIC LLC 01176
Encompass_Preferred
The module allows effortless attachment of chromatography columns. The bracket adjusts to the column circumference via a snap-in mechanism to provide a secure fit. For lengths greater than 15 cm of the suspended equipment, the use of two modules arranged one above the other is recommended to ensure the stability of the superstructure.
Catalog No. 50-255-8533
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.