Learn More
JDM SCIENTIFIC LLC Holder for glassware with ST14, extended
Supplier:  JDM SCIENTIFIC LLC 01113
Encompass_Preferred
The module corresponds in design and function to the module holder for glass devices with ST14. However, the extended version is particularly suitable for easily hanging particularly large round-bottomed flasks or separating funnels, for example.Note on joint tolerances for ST14:Maximum outside diameter at the narrowest point of the cone joint: 17.2 mm (larger diameters are too wide for the holder and cannot be held)Minimum outer diameter at the widest point of the cone joint: 18.0 mm (smaller diameters are too narrow for the holder and would fall out)
Catalog No. 50-255-8580
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.