Learn More
JDM SCIENTIFIC LLC Holder for glassware with ST29, plus closure
Supplier:  JDM SCIENTIFIC LLC 01115
Encompass_Preferred
The module corresponds to the module holder for glassware with ST29, but is also equipped with a rotable locking ring that protects glassware with standard joint ST29 against falling out.Note on joint tolerances for ST29:Maximum outside diameter at the narrowest point of the cone joint: 31.4 mm (larger diameters are too wide for the holder and cannot be held)Minimum outer diameter at the widest point of the cone joint: 32.8 mm (smaller diameters are too narrow for the holder and would fall out)
Catalog No. 50-255-8578
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.