Learn More
JDM SCIENTIFIC LLC Holder for pipettes
Supplier:  JDM SCIENTIFIC LLC 01145
Encompass_Preferred
The module allows easy attachment of up to three pipettes with hook. The pipette tips are not affected and therefore not contaminated.
Catalog No. 50-255-8562
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.