Learn More
JDM SCIENTIFIC LLC Holder for pipettes
Supplier:  JDM SCIENTIFIC LLC 01184
Encompass_Preferred
The module enables easy attachment of an INTEGRA VIAFLOW pipette or INTEGRA VOYAGER pipette. The pipette tips are not loaded and therefore not contaminated.
Catalog No. 50-255-8525
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.