Learn More
JDM SCIENTIFIC LLC Holder for tablet
Supplier:  JDM SCIENTIFIC LLC 01124
Encompass
The module allows easy attachment of tablets in portrait and landscape format. The tilted angle ensures good visibility and good usability of the display. The module can be adjusted to different tablet sizes via a locking mechanism, which can be adjusted in increments.
Catalog No. 50-255-8573
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.