Learn More
Medchemexpress LLC Homocarnosine TFA | 10MG
SDP
Supplier:  Medchemexpress LLC HY114883A10MG
Encompass_Preferred
Homocarnosine TFA is a dipeptide of -aminobutyric acid (GABA) and histidine unique to brain Homocarnosine TFA is an inhibitory neuromodulator synthesized in the neuron from GABA and exhibiting anticonvulsant effects[1] Homocarnosine TFA has antioxidant and anti-inflammatory actions prevention of DNA damage and inhibition of advanced glycation end-product formation[2]
Catalog No. 50-002-26383