Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Analytics™ GasAlertMicro 5 Series Gas Detector Power Accessories: Battery Pack, Black

Catalog No. 19045705
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
GasAlertMicro 5 Series Gas Detectors with Red Circuit Boards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-045-705 GasAlertMicro 5 Series Gas Detectors with Red Circuit Boards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-045-705 Supplier Honeywell Analytics™ Supplier No. M5BAT08BBP
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Color Black
For Use With (Equipment) GasAlertMicro 5 Series Gas Detectors with Red Circuit Boards
Type Lithium Polymer Battery Pack
Material Lithium polymer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.