Learn More
Honeywell Analytics™ GasAlertMicro 5 Series Gas Detector: Replacement Lamp
Catalog No. 19045709
Click to view available options
For Use With (Equipment):
PID Sensor (SR-Q07), GasAlertMicro 5 Series Gas Detectors
Catalog No. For Use With (Equipment)
19-045-709 PID Sensor (SR-Q07), GasAlertMicro 5 Series Gas Detectors
1 options
Catalog No. 19-045-709 Supplier Honeywell Analytics™ Supplier No. RLPID10.6BP
Specifications
For Use With (Equipment) PID Sensor (SR-Q07), GasAlertMicro 5 Series Gas Detectors
Type Replacement Lamp
Voltage 10.6V