Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Analytics™ ZoneGuard and GasChek Replacement Sensors

Catalog No. 19806787
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
ZoneGuard and Gas Chek Gas Detection Systems
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-806-787 ZoneGuard and Gas Chek Gas Detection Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-806-787 Supplier Honeywell Analytics™ Supplier No. 541633

missing translation for 'validMainframeMsg'

Specifications

Components Measured Ammonia
For Use With (Equipment) ZoneGuard and Gas Chek Gas Detection Systems
Type Replacement Sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.