Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Sperian™ Pure Fit™ 372 Lightweight Nitrile Coated Gloves

Catalog No. 19999480B
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-999-480B Medium
19-999-480C Large
19-999-480D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-999-480B Supplier Honeywell™ Sperian™ Supplier No. 372M
This item is not returnable. View return policy

Lightweight, 13-gauge nylon provides abrasion resistance while the nitrile coating protects against liquid penetration.

  • Gloves wear longer, and protect hands better than seamless knit cotton/polyester
  • Fully dipped gloves protect against liquid, and some chemical penetration
  • Pure Fit™ liners and coatings are lightweight and comfortable
  • Dexterity and tactile sensitivity for precision work
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Coated Area Full Glove
Construction Seamless Knit
Coating Material Nitrile
Color Black
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Dip-coated Gloves
Size Medium
For Use With (Application) Parts Assembly, Automotive Repairs, Painting, and Power Tool Operation
Glove Cuff Style Knit
Series 372
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.