Learn More
Honeywell™ North™ Silver Shield™ Chemical Resistant Aprons
Catalog No. 18999530 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Small
Universal
X-Large
Type:
Chemical-resistant Apron
Coat Apron
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Type
18-999-530 Universal Chemical-resistant Apron
19-088-1683D X-Large Coat Apron
19-150-0896 Small Coat Apron
19-150-0897 Medium Coat Apron
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-999-530 Supplier Honeywell™ Supplier No. SSA
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thin, lightweight aprons are available with and without sleeves.
  • Five layer PE and EVOH constructions provides excellent chemical resistant to over 280 different chemicals including alchohols, aliphatics, aromatice, chlorines, ketones and esters
  • Provides high performance protection in the most demanding applications
Specifications
Type Chemical-resistant Apron
Size Universal
Material Composite Fabric
Color Silver
Includes No Sleeves