Learn More
Honeywell™ Class 2 Traffic Safety Vest
Catalog No. 19120614
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-614 3X-Large
19-120-613 2X-Large
19-310-000 Large
19-310-001 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-120-614 Supplier Honeywell™ Supplier No. TVA2/15NZ/O3XL
This item is not returnable. View return policy
Enhances worker safety in hazardous situations where the visibility of the worker is a must, and nearby traffic exceeds speeds of 25 mph
  • Constructed from 100% polyester
  • Zipper front for quick and easy donning
  • Two 1 1/2 in. (3.8cm) North-Brite™ reflective vertical stripes on the front and back
  • One horizontal reflective stripe on the waist
Order Info
Available in orange or lime.
Compliance
Meets ANSI/ISEA 107, Class 2 standards.
TRUSTED_SUSTAINABILITY
Specifications
Color Orange
Size 3X-Large
Material Polyester