Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Knit Cotton Gloves with Dual Sided PVC-Dot Grip

Catalog No. 191500906
Change view
Click to view available options
Gender:
Women's
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gender
19-150-0906 Women's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-0906 Supplier Honeywell™ Supplier No. 82/6142S
This item is not returnable. View return policy

String knit shell is comfortable and helps keep hands clean in dirty applications, while PVC dots enhance grip capabilities and abrasion resistance.

  • PVC dot coating improves grip and provides abrasion resistance
  • String knit shell keeps hands clean in dirty applications
  • Knit wrist prevents dirt and debris from entering the glove
  • Reversible style can be worn on either hand
TRUSTED_SUSTAINABILITY

Specifications

Construction String Knit
Coating Material PVC Dots (2 Sides)
Color Natural
Hand Ambidextrous
Material Cotton
Size Universal
Style Reversible
For Use With (Application) General Handling and Maintenance
Gender Women's
Glove Cuff Style Knit
Series 82/6142
Length (English) 10 in.
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.