Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Eyesaline™ Personal Eyewash Bottle

Catalog No. 19085597 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Capacity (English):
4 oz.
8 oz.
32 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-085-597 32 oz.
19-003-027 4 oz.
19-811-412 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-085-597 Supplier Honeywell™ Supplier No. 320004550000H5
  • $286.00
    $240.65 / Case of 12

    Save $45.35

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Complete instructions for use

A Honewell Eyesaline™ Personal Eyewash Bottle provides immediate relief for eye injuries.

Carry-along squeeze bottles of saline solution.

  • Ideal for bathing eyes while transporting injured worker to hospital
  • Safety sealed and dated
  • Single-use only with snap-off tops for quick and easy access

Compliance

Does not meet the criteria of plumbed or self-contained eyewash equipment.

Specifications

Capacity (English) 32 oz.
Capacity (Metric) 946 mL
Certifications/Compliance FDA
For Use With (Application) First Aid Kit and Travel Kit, Agriculture, Chemical, Construction, Food Services, Oil and Gas, Steel, Welding Industries
Includes Buffered, Saline Solution and Superior to Tap Water
Material Polyethylene
Product Type Eyewash Bottle
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.