Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Eyesaline Personal Eyewash Bottles - Additional Sizes

Catalog No. 19003026 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Capacity (English):
1 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-003-026 1 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-003-026 Supplier Honeywell™ Supplier No. 320004510000
  • $213.00
    $183.65 / Case of 24

    Save $29.35

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Portable and readily available for use, these emergency eyewash bottles contain a buffered, saline solution that is sterile and superior to tap water for emergency.

Perfect for flushing nuisance particles such as pollen, dust, saw dust, smoke, etc., as well as for immediate treatment en route to primary eyewash unit or continued irrigation en route to medical care after a 15-minute eyewash.

  • Portable 1 oz bottles readily available for use
  • Bottles contain a buffered, saline solution that is sterile and superior to tap water for emergency eye care
  • Blow-fill-seal bottles are tamper resistant, yet easily removed in an emergency
  • User-control volume delivery delivers drops or an irrigating stream of solution
  • Ensured quality and lasting value with FDA-grade polyethylene bottles featuring lot numbers and expiration dates
  • 36-month shelf life from date of manufacturing
  • Personal eyewash devices are intended to supplement, not replace, primary eyewash units

Specifications

Capacity (English) 1 oz.
Capacity (Metric) 29.57 mL
Certifications/Compliance FDA
For Use With (Application) First Aid Kit and Travel Kit, Agriculture, Chemical, Construction, Food Services, Oil and Gas, Steel, Welding Industries
Includes Buffered, Saline Solution and Superior to Tap Water, Temper Proof Tab
Material Polyethylene
Product Type Eyewash Bottle
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.