Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ NorthFlex Light Task™ ESD Anti-Static Conductive Gloves

Catalog No. 19166718 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-166-718 2X-Large
19-166-713 X-Small
19-166-714 Small
19-166-715 Medium
19-166-716 Large
19-166-717 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-166-718 Supplier Honeywell™ North™ Supplier No. NF15ESD/11XXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lightweight, 15-gauge liner is constructed of copper based Thunderon™, with low surface resistivity when tested in accordance with EN 1149.

  • 15-gauge liner constructed of nylon and copper-based Thunderon™
  • Polyurethane coating on the palm and fingertips for enhanced grip capabilites
  • Low surface resistivity (<4 x 104 /sq.) when tested in accordance with EN 1149

Specifications

Certifications/Compliance ANSI/ISEA 105, ASTM D3389-05
Certifications/Compliance (Europe) EN 1149
Construction Seamless Knit
Coating Material Polyurethane
Color White
Exterior Finish Partially Textured
Hand Specific
Material Nylon, Thunderon
Product Type ESD Gloves
Size 2X-Large
For Use With (Application) Electronic Component Assembly/Handling
Glove Cuff Style Knit
Series NF15ESD
Yarn Gauge 15
Length (English) 20.5 in.
Length (Metric) 52 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.