Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ NorthFlex Light Task Plus 5™ Cut-Resistant Gloves

Catalog No. 19999128E Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-999-128E 2X-Large
19-999-128A Small
19-999-128B Medium
19-999-128C Large
19-999-128D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-999-128E Supplier Honeywell™ Supplier No. NFD20B/11XXL
  • $82.60
    $31.65 / Pair

    Save $50.95

    62% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $862.00
    $379.65 / Case of 12 PR

    Save $482.35

    56% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bi-polymer coated cut-resistant gloves is a blend that combines the best properties of nitrile and PU (polyurethane) to provide a surface that is mechanically tough and flexible.

  • Bi-polymer coating is a blend of nitrile and PU (polyurethane)
  • Comfortable, high performance cut-resistant
  • Seamless form fitting liner contains 'stretch-broken' Dynamo yarn that is cool and flexible
  • 13-gauge liner also contains glass fiber for enhanced cut resistance
  • Knit wrist

Specifications

Coating Material Bi-Polymer
Color Dark Blue
Cut Resistance Rating ANSI A5
Hand Ambidextrous
Latex Free Yes
Material Dyneema™
Product Type Cut-resistant Gloves
Size 2X-Large
For Use With (Application) Material handling and assembly work involving sharp objects like sheet metal, metal shavings, glass and metal parts
Glove Cuff Style Knit
Model NFD20B/11XXL
Series NorthFlex Light Task Plus 5™
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.