Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Perfect Fit™ 24 oz. Aramid Fiber Heat Resistant Gloves

Catalog No. 19050303
Change view
Click to view available options
Men's
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-050-303 Men's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-050-303 Supplier Honeywell™ Supplier No. KVT24ACKC

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Seamless knit gloves feature a Aramid Fiber exterior for heat/cut resistance, and a terrycloth interior for enhanced comfort.

  • 24 oz. Aramid Fiber loop-out exterior for cut/heat resistance
  • Terrycloth interior enhances heat protection and user comfort
  • Seamless knit construction eliminates seams that can rub or chafe
  • Continuous knit cuff provides wrist protection
TRUSTED_SUSTAINABILITY

Specifications

Construction Seamless Knit
Color Yellow, White
Cut Resistance Rating ANSI A4
For Use With (Application) Stamping, Sheet Metal, and Glass Handling
Gender Men's
Glove Cuff Style Continuous Knit
Series Perfect Fit™
Yarn Weight 24 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.