Learn More
Honeywell™ Hi Viz Tearaway Vest
Catalog No. 19120621
Change view
Click to view available options
Regular Size
X-Large Size
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-120-621 Regular Size
19-120-622 X-Large Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-120-621 Supplier Honeywell™ Supplier No. TV50RSTA
This item is not returnable. View return policy
Adds another measure of security for employees working around dangerous machinery or heavy traffic
  • Velcro™ closures on the back, front, shoulders and sides allow the vest to quickly pull away from the wearer if it becomes entangled with a passing vehicle or machinery. 100% polyester mesh vest is a hi-viz orange color
  • Pair of 2 in. (5.1cm) North-Brite™ reflective stripes
  • Stripes run parallel in the front and cross to form an “X” on the back
Order Info
Available in Regular or X-Large.
TRUSTED_SUSTAINABILITY
Specifications
Closure Type Hook and Loop Front
Color Orange
Size Standard
Material Polyester
Tape Type 1 in. Silver retroreflective vertical stripes