Learn More
Honeywell™ Lockout Stations
Catalog No. 19170138
Change view
Click to view available options
For Use With (Application):
Manufacturing, mining, municipal services, oil and gas, utilities
To withstand harsh conditions
Description:
Complete Four Lockout Station
Complete MT 10 Lockout Station
Complete MT Departmental Lockout Station
Complete Personal Lockout Station
Empty Four Lockout Station
Empty MT Departmental Lockout Station
Large Empty Lockout Station
Small Complete Lockout Station
Small Empty Lockout Station
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Description
19-170-138 To withstand harsh conditions Small Complete Lockout Station
19-170-139 To withstand harsh conditions Small Empty Lockout Station
19-170-140 To withstand harsh conditions Large Empty Lockout Station
19-170-143 Manufacturing, mining, municipal services, oil and gas, utilities Complete Personal Lockout Station
19-170-144 Manufacturing, mining, municipal services, oil and gas, utilities Empty Four Lockout Station
19-170-148 Manufacturing, mining, municipal services, oil and gas, utilities Empty MT Departmental Lockout Station
19-170-147 Manufacturing, mining, municipal services, oil and gas, utilities Complete MT 10 Lockout Station
19-170-145 Manufacturing, mining, municipal services, oil and gas, utilities Complete Four Lockout Station
19-170-149 Manufacturing, mining, municipal services, oil and gas, utilities Complete MT Departmental Lockout Station
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-170-138 Supplier Honeywell™ Supplier No. LSE101F
Specifications
Product Type Lockout Station
Material Plastic
Description Small Complete Lockout Station
For Use With (Application) To withstand harsh conditions
Includes LSE101 lockout panel, 2 in. VS02 V-Safe lockout, 4 in. VS04 V safe lockout, 6 in. VS06 V safe lockout, BS01 B safe lockout, LP110 E safe lockout 110 to 220V, LP550 E safe lockout 220 to 550V, MS86 M safe metal hasp, MS01 M safe nylon hasp
Size Small