Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Nitri-Kote™ Nitrile Coated Cotton/Polyester Gloves

Catalog No. 19050551B
Change view
Click to view available options
Size:
X-Small
Small
Medium
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-050-551B Small
19-150-0922 X-Small
19-050-551C Medium
19-150-0921 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-050-551B Supplier Honeywell™ North™ Supplier No. 78/1142S
This item is not returnable. View return policy

A unique coating process allows the glove to be fully coated for extended wear and breathability. The blended cotton/polyester shell reduces shrinking when laundered.

  • Ambidextrous design allows gloves to be replaced one at a time, rather than in pairs
  • Seamless construction increases comfort with no rubbing or chafing
  • Complies with USDA and FDA regulations (21 CFR) for use in food processing
  • Knit wrist prevents dirt and debris from entering the glove
TRUSTED_SUSTAINABILITY

Specifications

Coated Area Full Glove
Construction Seamless Knit
Coating Material Nitrile
Color Orange
Hand Ambidextrous
Material Polycotton
Size Small
For Use With (Application) Food Processing
Glove Cuff Style Knit
Series 78/1142
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.