Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Nomex™ SWAT Law Enforcement Gloves

Catalog No. 19170476
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-170-476 10
19-170-472 6
19-170-473 7
19-170-474 8
19-170-475 9
19-170-477 11
19-170-478 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-170-476 Supplier Honeywell™ Supplier No. NOMEXBLACK10

Constructed of 100% DuPont™ Nomex™ for protection against flash and flame hazards, with a sheepskin leather palm for enhanced grip capabilities. Gloves exceed all USAF specifications for thermal protection.

  • Constructed from 100% DuPont™ Nomex™ Grade A fibers for flame and heat resistance
  • Chrome-tanned, sheepskin leather palms enhance grip capabilities
  • Gloves exceed all USAF specifications for thermal protection
  • Ideal for law enforcement and/or SWAT applications

Compliance

Exceeds USAF Specifications for Thermal Protection

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Exceeds USAF Specifications for Thermal Protection
Color Black
Hand Specific
Material Nomex
Size 10
For Use With (Application) Law Enforcement and SWAT
Gender Unisex
Glove Cuff Style Open
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.