Learn More
Honeywell™ Nomex™ SWAT Law Enforcement Gloves
Description
- Constructed from 100% DuPont™ Nomex™ Grade A fibers for flame and heat resistance
- Chrome-tanned, sheepskin leather palms enhance grip capabilities
- Gloves exceed all USAF specifications for thermal protection
- Ideal for law enforcement and/or SWAT applications
Compliance
Exceeds USAF Specifications for Thermal Protection
Specifications
Specifications
| Certifications/Compliance | Exceeds USAF Specifications for Thermal Protection |
| Color | Black |
| Hand | Specific |
| Material | Nomex |
| Size | 11 |
| For Use With (Application) | Law Enforcement and SWAT |
| Gender | Unisex |
| Glove Cuff Style | Open |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.