Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Perfect-Coat Cut-Resistant Gloves

Catalog No. 19049642
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Hem Color:
Blue
Red
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hem Color
19-049-642 Medium Blue
19-049-641 Small Red
19-049-643 Large Yellow
19-049-644 X-Large White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-049-642 Supplier Honeywell™ Supplier No. KV303M
This item is not returnable. View return policy

Specifications

Coating Material Natural Rubber
Color Yellow
Cut Resistance Rating ANSI A4
Material Aramid Fiber, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Protects against cuts and abrasions from glass, metals and other sharp objects
Glove Cuff Color Blue
Glove Cuff Style Knit
Hem Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.