Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Perfect-Coat™ KV350 Aramid Fiber Cut-Resistant Gloves, Nitrile Coated

Catalog No. 19816557
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-816-557 Medium
19-816-556 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-816-557 Supplier Honeywell™ Supplier No. KV350M

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Medium weight seamless knit liner provides cut and abrasion protection in hazardous applications, while nitrile coating on the palm and fingertips enhances grip capabilities.

  • Aramid fiber is 5x stronger than steel on an equal weight basis
  • Gloves provide cut and abrasion protection in hazardous applications
  • 10-gauge medium-weight seamless knit liner
  • Nitrile coating on the palm and fingertips
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Construction Seamless Knit
Coating Material Nitrile
Color Yellow, Green
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Hand Specific
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Automotive, mechanical, fabrication, glass handling, metal stamping, sheet metal, recycling
Glove Cuff Style Knit
Model KV350-M
Series KV350
Yarn Gauge 10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.