Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Perfect Fit KV13 Aramid Fiber Gloves

Catalog No. 19816549
Change view
Click to view available options
Size:
Jumbo
Universal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-816-549 Universal
19-816-550 Jumbo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-816-549 Supplier Honeywell™ Supplier No. KV13A
This item is not returnable. View return policy

Seamless knit gloves of aramid fibers have a strength which is five times stronger than steel on an equal weight basis.

  • Aramid fiber
  • Seamless knit
  • 13-gauge
  • Level 2 ANSI ASTM cut protection
  • 13-gauge
  • Yellow, lightweight, uncoated
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 1
Construction Seamless Knit
Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A2
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Universal
For Use With (Application) Light duty applications
Gender Men's
Model KV13A
Series Perfect Fit
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.