Learn More
Honeywell™ PowerCoat PVC Chemical Resistant Gloves, Thermal Fleece Lining
Description
- PVC coating protects hands in hazardous liquid applications
- Coating is soft and comfortable
- Flexibility reduces hand fatigue
- Thermal fleece lining keeps hands warm and provides a cushion for lifting heavy items
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 511, EN 374-3 |
| Color | Orange |
| Latex Free | Yes |
| Material | PVC |
| Size | Large |
| Thickness (English) | 137 mil |
| Thickness (Metric) | 3.5 mm |
| EN388 Standard | EN 388:4121 |
| EN ISO 374-1 CHEMICAL LETTER CODES | AJL |
| Glove Cuff Style | Rolled |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.