Learn More
Honeywell™ PRO 3003 Leather Station/Duty Boots, Wide Width
Extremely lightweight and comfortable
Supplier: Honeywell™ BT30030703
Description
- Constructed of water-resistant military AB leather for station/duty use or any use where a NFPA standard is not required
- Leather lace up/seed zip
- Cambrelle™ liner wicks away moisture and adds comfort
- Obilque-shaped safety toe cap follows natural contour of foot for greater comfort
- Full wrap-around Goodyear welt adds support and reduces fatigue and ankle strains and sprains
- Nonmarking Vibram™ lug sole
- Thermoplastic backpart molded heel counter maintains shape in moist conditions
- Triple linear ridge shank for increased arch support
- Puncture-resistant steel bottom plate
- 8 in. high
Specifications
8 in. | |
Cambrelle | |
Leather | |
7E |