Learn More
Honeywell™ PRO 3003 Leather Station/Duty Boots, Wide Width
Extremely lightweight and comfortable
Supplier:  Honeywell™ BT30031403
 View more versions of this product
Catalog No. 19-168-846
Only null left
Add to Cart
Description
Description
  • Constructed of water-resistant military AB leather for station/duty use or any use where a NFPA standard is not required
  • Leather lace up/seed zip
  • Cambrelle™ liner wicks away moisture and adds comfort
  • Obilque-shaped safety toe cap follows natural contour of foot for greater comfort
  • Full wrap-around Goodyear welt adds support and reduces fatigue and ankle strains and sprains
  • Nonmarking Vibram™ lug sole
  • Thermoplastic backpart molded heel counter maintains shape in moist conditions
  • Triple linear ridge shank for increased arch support
  • Puncture-resistant steel bottom plate
  • 8 in. high
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8 in.
Cambrelle
Leather
14E
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.