Learn More
Honeywell™ PRO 3003 Leather Station/Duty Boots, Wide Width
Catalog No. 19168848
Change view
Click to view available options
Size:
10.5E
10.5EEE
10E
10EEE
11.5E
11.5EEE
11E
11EEE
12E
12EEE
13E
13EEE
14E
14EEE
6.5E
6E
7.5E
7.5EEE
7E
7EEE
8.5E
8.5EEE
8E
8EEE
9.5E
9.5EEE
9E
9EEE
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-168-848 7.5EEE
19-168-853 10EEE
19-168-843 11.5E
19-168-854 11EEE
19-168-855 11.5EEE
19-168-842 11E
19-823-060 10.5EEE
19-168-844 12E
19-168-846 14E
19-168-845 13E
19-168-858 14EEE
19-168-856 12EEE
19-168-857 13EEE
19-168-834 6E
19-168-835 6.5E
19-168-836 7E
19-168-847 7EEE
19-168-837 7.5E
19-168-839 8.5E
19-168-838 8E
19-168-840 9E
19-168-852 9.5EEE
19-168-850 8.5EEE
19-168-849 8EEE
19-168-841 9.5E
19-168-851 9EEE
19-814-338 10E
19-823-059 10.5E
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 missing translation for 'options'
Catalog No. 19-168-848 missing translation for 'mfr' Honeywell™ missing translation for 'supplierNo' BT30030755
Add to Cart
Add to Cart
Extremely lightweight and comfortable
  • Constructed of water-resistant military AB leather for station/duty use or any use where a NFPA standard is not required
  • Leather lace up/seed zip
  • Cambrelle™ liner wicks away moisture and adds comfort
  • Obilque-shaped safety toe cap follows natural contour of foot for greater comfort
  • Full wrap-around Goodyear welt adds support and reduces fatigue and ankle strains and sprains
  • Nonmarking Vibram™ lug sole
  • Thermoplastic backpart molded heel counter maintains shape in moist conditions
  • Triple linear ridge shank for increased arch support
  • Puncture-resistant steel bottom plate
  • 8 in. high
TRUSTED_SUSTAINABILITY
Specifications
Height (English) 8 in.
Liner Material Cambrelle
Material Leather
Size 7.5EEE