Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Pure Fit™ PF541 Cut-Resistant Gloves

Catalog No. 19148402
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Glove Cuff Color:
Blue
Red
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-148-402 Large Yellow
19-148-400 Small Red
19-148-401 Medium Blue
19-148-403 X-Large White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-148-402 Supplier Honeywell™ Supplier No. PF541L
This item is not returnable. View return policy

Lightweight gloves with polyurethane coating are ideal for cut and puncture protection without sacrificing dexterity.

  • Black and white speckled shell made of low-linting HPPE
  • Gray polyurethane smooth coating of palm and fingertip provides added grip, abrasion resistance and tactile sensitivity
  • Gray color of gloves hides dirt and grime
  • Glove cuff color differentiates sizing

TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Color Black, Gray, Salt and Pepper
Cut Resistance Rating ANSI A3
Material HPPE, Lycra
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388:4342
For Use With (Application) Provides added grip, abrasion resistance and tactile sensitivity
Glove Cuff Color Yellow
Glove Cuff Style Knit
Hem Color Yellow
Model PF541-L
Series Pure Fit™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.