Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Pure Fit™ 380 Nitrile Coated Nylon Gloves

Catalog No. 19121740C
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-121-740C Large
19-121-740B Medium
19-121-740D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-121-740C Supplier Honeywell™ Supplier No. 380L

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

Nitrile coating on the palm reduces hand fatigue, while also enhancing grip capabilities. Seamless knit nylon liner is comfortable, and minimizes irritation.

  • Constructed of lightweight, seamless knit nylon
  • Dipped palm and fingertips improve dexterity, and reduce hand fatigue
  • Coating provides an extra wear area for extended use, and increased comfort
  • Low linting for cleanroom applications
TRUSTED_SUSTAINABILITY

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile
Color Black
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Parts Assembly/Handling, Automotive Assembly, Painting, and Power Tool Operation
Glove Cuff Style Knit
Series Pure Fit™ 380
Yarn Gauge 13
Show More Show Less

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.