Learn More
Honeywell™ Pure Fit™ HPPE Cut-Resistant Gloves, Polyurethane Coated
Description
- Lightweight Dyneema fabric with polyurethane palm and fingertip coating for increased cut, slash and abrasion resistance
- Low-lint for clean applications
- Suited for use in electronic and small parts assembly, high-tech applications, automotive subassembly and inspection
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 6 |
| Color | Gray |
| Cut Resistance Rating | ANSI A1 |
| Material | HPPE, Lycra |
| Product Type | Cut-resistant Gloves |
| Size | Medium |
| EN388 Standard | EN 388 |
| For Use With (Application) | electronic/small parts assembly, high tech applications, automotive sub assembly, light fabrication, micro engineering, packaging, inspection and light industrial |
| Glove Cuff Color | Blue |
| Hem Color | Blue |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.