Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Pure Fit Polyurethane-Coated Cut Resistant Nylon Gloves

Catalog No. 191201214
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-1214 X-Large
19-120-1212 Medium
19-120-1213 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-120-1214 Supplier Honeywell™ Supplier No. PF550XL

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

Lightweight gloves feature polyurethane coating on the palm and fingers for abrasion/cut resistance, and enhanced grip capabilities.

  • Constructed from low-linting nylon with a high tensile strength
  • Ideal for small precision work where dexterity and tactile sensitivity are crucial
  • Polyurethane coating provides abrasion resistance and enhanced grip capabilities
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A1
Hand Specific
Material Nylon
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Small Parts Assembly, Automotive Repair, Metal Handling and Painting
Glove Cuff Color Black
Glove Cuff Style Knit
Model PF550-XL
Series Pure Fit 550
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.